Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001221.3 | NP_001212.2 | 13 - 14 | 1845 | 92 | |
NM_172115.2 | NP_742113.1 | 13 - 14 | 1845 | 92 | ||
NM_172127.2 | NP_742125.1 | 13 - 14 | 1845 | 92 | ||
NM_172128.2 | NP_742126.1 | 13 - 14 | 1845 | 92 | ||
XM_006714330.2 | XP_006714393.1 | 13 - 14 | 1684 | 92 | ||
GenBank mRNA | AF071569.1 | - | 13 - 14 | 986 | 92 | |
AK025368.1 | - | 2 - 3 | 96 | 92 | ||
AK055642.1 | - | 13 - 14 | 1363 | 92 | ||
BC032784.1 | - | 13 - 14 | 1048 | 5178265 | 92 | |
U50361.1 | - | 2 - 3 | 83 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons |