Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008093.hg.1 |
ENST00000307518.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008093.hg.1 |
ENST00000370655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008093.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008093.hg.1 |
TC10000698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008580.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000298808.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000307518.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000370655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
ENST00000455090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001129981.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001291218.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_001291219.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
NM_020349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158503.hg.1 |
TC1000008580.hg.1 |
Clariom D TC |