Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318192.1 | NP_001305121.1 | 15 - 16 | 2455 | 80 | |
NM_012450.3 | NP_036582.2 | 15 - 16 | 2452 | 80 | ||
XM_017011962.1 | XP_016867451.1 | 13 - 14 | 1823 | 80 | ||
GenBank mRNA | AB208809.1 | - | 12 - 13 | 6730 | 80 | |
AF169301.1 | - | 15 - 16 | 2425 | 80 | ||
AK313072.1 | - | 15 - 16 | 2045 | 80 | ||
BC030689.2 | - | 15 - 16 | 2434 | 4821193 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |