Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061528.hg.1 |
TC01004095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061528.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061528.hg.1 |
ENST00000411742.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061528.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01061528.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236392.hg.1 |
TC0100018142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000366472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000475978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000484202.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
ENST00000494837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018142.hg.1 |
NM_030645.1 |
Refseq |