Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029612.hg.1 |
TC07001909.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029612.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029612.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012760.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191986.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191986.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191986.hg.1 |
ENST00000275766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191986.hg.1 |
NM_080660.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191986.hg.1 |
TC0700012760.hg.1 |
Clariom D TC |