Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013197.hg.1 |
ENST00000353258.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013197.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16013197.hg.1 |
TC16000918.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000563645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009642.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000353258.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000564172.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000569773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
ENST00000569899.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
NM_016641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600173753.hg.1 |
TC1600009642.hg.1 |
Clariom D TC |