Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
ENST00000324460.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011523.hg.1 |
NM_005347.4 |
Refseq |