Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022727.hg.1 |
TC07001509.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022727.hg.1 |
ENST00000055077.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022727.hg.1 |
ENST00000352131.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022727.hg.1 |
NM_002914.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07022727.hg.1 |
NM_181471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000055077.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000352131.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000463194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000470266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000479105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000480432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000491206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000493156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000494019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000497430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000621097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_002914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_181471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000055077.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000352131.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000463194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000470266.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000479105.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000480432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000491206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000493156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000494019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000497430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
ENST00000621097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278792.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_001278793.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_002914.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
NM_181471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700181837.hg.1 |
TC0700011510.hg.1 |
Clariom D TC |