Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007242.2 | NP_001007243.2 | 2 - 3 | 283 | 71 | |
GenBank mRNA | AB035542.1 | - | 1 - 2 | 72 | 71 | |
BC032693.2 | - | 1 - 2 | 130 | 5229838 | 71 | |
BG661274.1 | - | 1 - 2 | 130 | 71 | ||
BX648201.1 | - | 2 - 3 | 170 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |