Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014246.1 | NP_055061.1 | 30 - 31 | 8299 | 95 | |
XM_006724383.3 | XP_006724446.1 | 30 - 31 | 8626 | 95 | ||
XM_011530554.2 | XP_011528856.1 | 30 - 31 | 4842 | 95 | ||
XM_011530555.2 | XP_011528857.1 | 30 - 31 | 4923 | 95 | ||
GenBank mRNA | AF231024.1 | - | 30 - 31 | 8299 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |