Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169122.1 | NP_001162593.1 | 12 - 13 | 3290 | 82 | |
GenBank mRNA | L76569.1 | - | 12 - 13 | 3203 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |