Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUWE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031407.6 | NP_113584.3 | 39 - 40 | 5224 | 100 | |
XM_005261965.3 | XP_005262022.1 | 38 - 39 | 5291 | 100 | ||
XM_011530747.1 | XP_011529049.1 | 40 - 41 | 5467 | 100 | ||
XM_011530751.2 | XP_011529053.1 | 39 - 40 | 5335 | 100 | ||
XM_017029191.1 | XP_016884680.1 | 38 - 39 | 8219 | 100 | ||
XM_017029192.1 | XP_016884681.1 | 38 - 39 | 8219 | 100 | ||
XM_017029193.1 | XP_016884682.1 | 38 - 39 | 8198 | 100 | ||
XM_017029194.1 | XP_016884683.1 | 38 - 39 | 8219 | 100 | ||
XM_017029195.1 | XP_016884684.1 | 38 - 39 | 8219 | 100 | ||
XM_017029196.1 | XP_016884685.1 | 38 - 39 | 8219 | 100 | ||
XM_017029197.1 | XP_016884686.1 | 38 - 39 | 8219 | 100 | ||
XM_017029198.1 | XP_016884687.1 | 39 - 40 | 5561 | 100 | ||
XM_017029199.1 | XP_016884688.1 | 40 - 41 | 5488 | 100 | ||
XM_017029200.1 | XP_016884689.1 | 39 - 40 | 5358 | 100 | ||
XM_017029201.1 | XP_016884690.1 | 39 - 40 | 5375 | 100 | ||
XM_017029202.1 | XP_016884691.1 | 39 - 40 | 5351 | 100 | ||
XM_017029203.1 | XP_016884692.1 | 39 - 40 | 5357 | 100 | ||
XM_017029204.1 | XP_016884693.1 | 37 - 38 | 7969 | 100 | ||
XM_017029205.1 | XP_016884694.1 | 37 - 38 | 7949 | 100 | ||
XM_017029206.1 | XP_016884695.1 | 38 - 39 | 8219 | 100 | ||
GenBank mRNA | AB002310.3 | - | 8 - 9 | 1326 | 100 | |
AB071605.1 | - | 12 - 13 | 1924 | 100 | ||
AK056671.1 | - | 22 - 23 | 3450 | 100 | ||
AK091352.1 | - | 11 - 12 | 1742 | 100 | ||
AK294575.1 | - | 26 - 27 | 3824 | 100 | ||
AY772009.1 | - | 37 - 38 | 4869 | 100 | ||
AY929612.1 | - | 36 - 37 | 4821 | 100 | ||
DQ097177.1 | - | 39 - 40 | 5223 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |