Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014358.3 | NP_055173.1 | 3 - 4 | 387 | 116 | |
XM_011520614.2 | XP_011518916.1 | 2 - 3 | 660 | 116 | ||
GenBank mRNA | AB024718.1 | - | 3 - 4 | 373 | 116 | |
AK312671.1 | - | 3 - 4 | 388 | 116 | ||
AY358499.1 | - | 3 - 4 | 327 | 116 | ||
BC000715.2 | - | 3 - 4 | 305 | 3507103 | 116 | |
DA678013.1 | - | 3 - 4 | 388 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |