Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000261168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000536444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000540793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000541654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000543189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
ENST00000544627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
NM_018179.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002419.hg.1 |
TC12000193.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000261168.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000536444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000537653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000538511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000539659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000540793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000541654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000543189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000544627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_001286514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_001286515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_018179.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_181352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000261168.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000536444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000537653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000538511.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000539659.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000540793.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000541654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000543189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
ENST00000544627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_001286514.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_001286515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_018179.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
NM_181352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148475.hg.1 |
TC1200006935.hg.1 |
Clariom D TC |