Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
ENST00000342386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
ENST00000375940.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
ENST00000375946.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002242.hg.1 |
TC10000189.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
ENST00000477034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007092.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000342386.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000375940.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
ENST00000375946.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_001172303.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_001172304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
NM_032844.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000148388.hg.1 |
TC1000007092.hg.1 |
Clariom D TC |