Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304331.1 | NP_001291260.1 | 23 - 24 | 3356 | 112 | |
XM_006711586.2 | XP_006711649.1 | 23 - 24 | 3386 | 112 | ||
XM_011510070.2 | XP_011508372.1 | 11 - 12 | 1415 | 112 | ||
XM_017002607.1 | XP_016858096.1 | 22 - 23 | 3251 | 112 | ||
XM_017002609.1 | XP_016858098.1 | 22 - 23 | 3251 | 112 | ||
GenBank mRNA | AB020704.1 | - | 5 - 6 | 485 | 112 | |
AK126624.1 | - | 10 - 11 | 1924 | 112 | ||
AK295757.1 | - | 17 - 18 | 2084 | 112 | ||
BC132923.1 | - | 10 - 11 | 1866 | 40146945 | 112 | |
BC132925.1 | - | 10 - 11 | 1866 | 40146947 | 112 | |
EF428334.1 | - | 28 - 29 | 3223 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |