Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
TC01003501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000333360.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367779.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367781.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
ENST00000367782.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054450.hg.1 |
NM_000450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000333360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
NM_000450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000333360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367775.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
ENST00000367777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
NM_000450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244031.hg.1 |
TC0100018529.hg.1 |
Clariom D TC |