Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012727.hg.1 |
TC07000809.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012727.hg.1 |
ENST00000222481.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012727.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164945.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164945.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164945.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164945.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164945.hg.1 |
TC0700009132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000222481.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000416698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000480781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
ENST00000487259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009132.hg.1 |
NM_001869.2 |
Refseq |