Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000092.hg.1 |
ENST00000166139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000092.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000092.hg.1 |
TC19000012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144354.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144354.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144354.hg.1 |
TC1900006477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144354.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144354.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000588773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000591573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000592947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
ENST00000605925.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006477.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
NM_005860.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
TC1900006477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000166139.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000589185.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
ENST00000592058.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144357.hg.1 |
NM_005860.2 |
Refseq |