Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU517121.1 | - | 1 - 2 | 137 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |