Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007284.hg.1 |
ENST00000338437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007284.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007284.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007284.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007284.hg.1 |
TC16000522.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600159966.hg.1 |
TC1600008134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000338437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000394069.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
ENST00000548332.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_001014435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008134.hg.1 |
NM_005182.2 |
Refseq |