Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000488234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000488234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032473.hg.1 |
TC02002033.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032473.hg.1 |
ENST00000393868.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032473.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196699.hg.1 |
TC0200013232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000393868.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000430989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000442240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000483605.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000484365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000487809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000488234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000488234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
ENST00000491642.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013232.hg.1 |
NM_003849.3 |
Refseq |