Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006073.3 | NP_006064.2 | 31 - 32 | 2060 | 89 | |
GenBank mRNA | U18985.1 | - | 31 - 32 | 1764 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |