Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005947.2 | NP_005938.1 | 1 - 2 | 82 | 134 | |
GenBank mRNA | AF349000.2 | - | 1 - 2 | 81 | 134 | |
AY168638.1 | - | 1 - 2 | 83 | 134 | ||
BC069421.1 | - | 1 - 2 | 77 | 7262703 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |