Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005266.hg.1 |
TC09000490.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005266.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005266.hg.1 |
ENST00000422139.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09005266.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154640.hg.1 |
TC0900008148.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000355295.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
ENST00000492428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_001302884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008148.hg.1 |
NM_014290.2 |
Refseq |