Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
TC01001907.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
ENST00000541865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
ENST00000543291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
ENST00000543760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029613.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000488903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000488903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000485438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000488903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000488903.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000492568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011920.hg.1 |
NR_120373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
ENST00000366672.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
ENST00000494106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
NM_001291866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
NM_004481.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100187962.hg.1 |
TC0100011920.hg.1 |
Clariom D TC |