Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
TC02000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000282412.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000345249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000378540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000378551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000409432.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
ENST00000409895.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
NM_001033557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
NM_002706.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
NM_177968.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004084.hg.1 |
NM_177969.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
TC02000274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000282412.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000345249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000378540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000378551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000409432.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
ENST00000409895.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
NM_001033557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
NM_002706.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
NM_177968.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02004085.hg.1 |
NM_177969.3 |
Refseq |