Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017017728.1 | XP_016873217.1 | 3 - 4 | 422 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |