Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
ENST00000305253.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
ENST00000534099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002196.hg.1 |
TC11000155.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002195.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002195.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11002195.hg.1 |
TC11000155.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100147509.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147509.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147509.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147509.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147509.hg.1 |
TC1100006773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147511.hg.1 |
TC1100006773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147510.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147510.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147510.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147510.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147510.hg.1 |
TC1100006773.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000299506.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000305253.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
ENST00000534099.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_003320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006773.hg.1 |
NM_177972.2 |
Refseq |