Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017539.2 | NP_060009.1 | 52 - 53 | 8455 | 106 | |
XM_006721062.1 | XP_006721125.1 | 52 - 53 | 8593 | 106 | ||
XM_011545882.2 | XP_011544184.1 | 53 - 54 | 9033 | 106 | ||
XM_011545883.1 | XP_011544185.1 | 53 - 54 | 8801 | 106 | ||
XM_011545885.2 | XP_011544187.1 | 53 - 54 | 9033 | 106 | ||
XM_011545886.2 | XP_011544188.1 | 45 - 46 | 7408 | 106 | ||
XM_011545888.2 | XP_011544190.1 | 35 - 36 | 5852 | 106 | ||
XM_011545889.2 | XP_011544191.1 | 35 - 36 | 5816 | 106 | ||
XM_017023426.1 | XP_016878915.1 | 52 - 53 | 8898 | 106 | ||
XM_017023427.1 | XP_016878916.1 | 51 - 52 | 8790 | 106 | ||
XM_017023428.1 | XP_016878917.1 | 50 - 51 | 8454 | 106 | ||
XM_017023430.1 | XP_016878919.1 | 11 - 12 | 2475 | 106 | ||
XM_017023432.1 | XP_016878921.1 | 52 - 53 | 8593 | 106 | ||
GenBank mRNA | AF494040.1 | - | 52 - 53 | 8455 | 106 | |
AK125952.1 | - | 5 - 6 | 808 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |