Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006765.3 | NP_006756.2 | 9 - 10 | 1372 | 70 | |
XM_011544652.2 | XP_011542954.1 | 8 - 9 | 915 | 70 | ||
XM_017013861.1 | XP_016869350.1 | 8 - 9 | 915 | 70 | ||
GenBank mRNA | AY562998.1 | - | 2 - 3 | 131 | 70 | |
U42349.1 | - | 9 - 10 | 1185 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |