Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HMBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324383.1 | NP_001311312.1 | 10 - 11 | 1556 | 72 | |
NM_001324384.1 | NP_001311313.1 | 10 - 11 | 1559 | 72 | ||
NM_001324385.1 | NP_001311314.1 | 10 - 11 | 1550 | 72 | ||
XM_005273635.1 | XP_005273692.1 | 10 - 11 | 1529 | 72 | ||
XM_017013823.1 | XP_016869312.1 | 10 - 11 | 1564 | 72 | ||
GenBank mRNA | AK295320.1 | - | 9 - 10 | 1192 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons |