Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000354336.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
ENST00000411769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006681.hg.1 |
NM_005207.3 |
Refseq |