Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019422.hg.1 |
TC17002915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019422.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019422.hg.1 |
ENST00000394042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019422.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019423.hg.1 |
TC17002915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019423.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019423.hg.1 |
ENST00000394042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019423.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019424.hg.1 |
TC17002915.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019424.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019424.hg.1 |
ENST00000394042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019424.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206259.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206259.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206259.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206259.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206259.hg.1 |
TC1700012397.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700206258.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206258.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206258.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206258.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206258.hg.1 |
TC1700012397.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012397.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206261.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206261.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206261.hg.1 |
ENST00000312150.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206261.hg.1 |
NM_181534.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206261.hg.1 |
TC1700012397.hg.1 |
Clariom D TC |