Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006980.hg.1 |
TC07000495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006980.hg.1 |
ENST00000415482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006980.hg.1 |
ENST00000543073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006980.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006980.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
TC07000495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
ENST00000415482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
ENST00000435495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
ENST00000543073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006981.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
TC07000495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
ENST00000248594.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
ENST00000415482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
ENST00000435495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
ENST00000543073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006987.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
TC07000495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
ENST00000248594.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
ENST00000415482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
ENST00000435495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
ENST00000543073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006989.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
TC07000495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
ENST00000248594.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
ENST00000415482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
ENST00000435495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
ENST00000543073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07006988.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156504.hg.1 |
TC0700008141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156515.hg.1 |
TC0700008141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000523952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
ENST00000523952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156514.hg.1 |
TC0700008141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156503.hg.1 |
TC0700008141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000440186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000523952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000248594.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000415482.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000418110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000433369.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000435495.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000440186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000460731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000522115.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
ENST00000523952.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_001131008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_001131009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
NM_002835.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156511.hg.1 |
TC0700008141.hg.1 |
Clariom D TC |