Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010189.hg.1 |
TC06000870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010189.hg.1 |
ENST00000368972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010189.hg.1 |
ENST00000392644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010189.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600162307.hg.1 |
TC0600009076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000237512.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000368972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000392644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000414610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
ENST00000481850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_001286609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009076.hg.1 |
NM_032131.5 |
Refseq |