Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014611.2 | NP_055426.1 | 15 - 16 | 2332 | 103 | |
XM_011535635.2 | XP_011533937.1 | 15 - 16 | 2137 | 103 | ||
XM_011535636.2 | XP_011533938.1 | 15 - 16 | 2302 | 103 | ||
GenBank mRNA | AF503925.1 | - | 15 - 16 | 2261 | 103 | |
AK098453.1 | - | 7 - 8 | 805 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |