Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001646.hg.1 |
ENST00000379487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001646.hg.1 |
ENST00000542082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001646.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001646.hg.1 |
TC13000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001647.hg.1 |
ENST00000379487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001647.hg.1 |
ENST00000542082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001647.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001647.hg.1 |
TC13000141.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300147232.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147232.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147232.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147232.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147232.hg.1 |
TC1300006965.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006965.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147233.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147233.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147233.hg.1 |
ENST00000379487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147233.hg.1 |
NM_007187.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147233.hg.1 |
TC1300006965.hg.1 |
Clariom D TC |