Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008079.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008079.hg.1 |
TC20000558.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
ENST00000539905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
ENST00000540134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008076.hg.1 |
TC20000558.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000157590.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157590.hg.1 |
TC2000008175.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157590.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008175.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_024411.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
TC2000008175.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000217305.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000539905.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
ENST00000540134.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190899.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_001190900.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000157586.hg.1 |
NM_024411.4 |
Refseq |