Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018961.3 | NP_061834.1 | 5 - 6 | 755 | 100 | |
XM_006724013.3 | XP_006724076.1 | 5 - 6 | 839 | 100 | ||
XM_011529605.2 | XP_011527907.1 | 5 - 6 | 755 | 100 | ||
XM_011529606.2 | XP_011527908.1 | 5 - 6 | 724 | 100 | ||
XM_011529607.2 | XP_011527909.1 | 5 - 6 | 808 | 100 | ||
XM_011529609.1 | XP_011527911.1 | 5 - 6 | 755 | 100 | ||
GenBank mRNA | AF520809.1 | - | 5 - 6 | 682 | 100 | |
AJ277750.1 | - | 5 - 6 | 702 | 100 | ||
BC069357.1 | - | 5 - 6 | 702 | 7262315 | 100 | |
BC069483.1 | - | 5 - 6 | 702 | 7262391 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |