Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021436.hg.1 |
ENST00000222032.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021436.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021436.hg.1 |
TC19001572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021438.hg.1 |
ENST00000222032.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021438.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021438.hg.1 |
TC19001572.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
TC1900010801.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189386.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010801.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
NM_032488.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
TC1900010801.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
ENST00000222032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
ENST00000597255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900189383.hg.1 |
NM_032488.3 |
Refseq |