Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000882.hg.1 |
ENST00000379215.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000882.hg.1 |
ENST00000496136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000882.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000882.hg.1 |
TC10000086.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000885.hg.1 |
ENST00000379215.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000885.hg.1 |
ENST00000496136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000885.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000885.hg.1 |
TC10000086.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000880.hg.1 |
ENST00000379215.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000880.hg.1 |
ENST00000496136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000880.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000880.hg.1 |
TC10000086.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146293.hg.1 |
TC1000006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146291.hg.1 |
TC1000006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000495787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006782.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146294.hg.1 |
TC1000006782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000379215.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000420401.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000422887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
ENST00000496136.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
NM_024693.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146297.hg.1 |
TC1000006782.hg.1 |
Clariom D TC |