Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010963.hg.1 |
ENST00000280245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010963.hg.1 |
ENST00000342404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010963.hg.1 |
ENST00000376067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010963.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010963.hg.1 |
TC11000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010962.hg.1 |
ENST00000280245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010962.hg.1 |
ENST00000376067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010962.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010962.hg.1 |
TC11000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010964.hg.1 |
ENST00000280245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010964.hg.1 |
ENST00000342404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010964.hg.1 |
ENST00000376067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010964.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010964.hg.1 |
TC11000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010965.hg.1 |
ENST00000280245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010965.hg.1 |
ENST00000342404.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010965.hg.1 |
ENST00000376067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010965.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010965.hg.1 |
TC11000866.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163996.hg.1 |
TC1100008653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163993.hg.1 |
TC1100008653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163995.hg.1 |
TC1100008653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000526729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
ENST00000529676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008653.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163992.hg.1 |
TC1100008653.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
ENST00000280245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
ENST00000342404.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
NM_001286159.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
NM_173556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100163994.hg.1 |
TC1100008653.hg.1 |
Clariom D TC |