Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001026.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001026.hg.1 |
ENST00000441671.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001026.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001026.hg.1 |
TC12000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001024.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001024.hg.1 |
ENST00000441671.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001024.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001024.hg.1 |
TC12000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
ENST00000541049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006644.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145945.hg.1 |
TC1200006644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145942.hg.1 |
TC1200006644.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000203629.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000441671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
ENST00000538079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
NM_002286.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145943.hg.1 |
TC1200006644.hg.1 |
Clariom D TC |