Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003370.hg.1 |
ENST00000221486.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003370.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003370.hg.1 |
TC19000229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
TC1900007098.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900150994.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000221486.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590121.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000590279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
ENST00000593017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007098.hg.1 |
NM_006397.2 |
Refseq |