Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010221.hg.1 |
TC05000717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010221.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010221.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010220.hg.1 |
TC05000717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010220.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010220.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010219.hg.1 |
TC05000717.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010219.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05010219.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161074.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161074.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161074.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161074.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161074.hg.1 |
TC0500008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161072.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161072.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161072.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161072.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161072.hg.1 |
TC0500008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500161070.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161070.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161070.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500161070.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500161070.hg.1 |
TC0500008842.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
ENST00000274710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008842.hg.1 |
NM_032289.3 |
Refseq |