Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00959280_m1
See other GRB10 GE Assays ›
Gene Symbol:
GRB10
Gene Name:
growth factor receptor bound protein 10
Gene Aliases:
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location:
Chr.7: 50590063 - 50793462 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00959280_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: GRB10
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001001550.2
NP_001001550.1
8 - 9
1096
95
NM_001001555.2
NP_001001555.1
9 - 10
1343
95
NM_005311.4
NP_005302.3
8 - 9
1016
95
XM_011515304.1
XP_011513606.1
11 - 12
1455
95
XM_011515312.2
XP_011513614.1
10 - 11
1743
95
XM_011515316.1
XP_011513618.1
9 - 10
1288
95
XM_011515323.2
XP_011513625.1
6 - 7
2792
95
XM_017012029.1
XP_016867518.1
10 - 11
1675
95
XM_017012030.1
XP_016867519.1
9 - 10
1609
95
XM_017012031.1
XP_016867520.1
9 - 10
1540
95
XM_017012032.1
XP_016867521.1
15 - 16
2052
95
XM_017012033.1
XP_016867522.1
12 - 13
1590
95
XM_017012034.1
XP_016867523.1
10 - 11
1191
95
XM_017012035.1
XP_016867524.1
16 - 17
3101
95
XM_017012036.1
XP_016867525.1
17 - 18
2886
95
XM_017012037.1
XP_016867526.1
17 - 18
2723
95
XM_017012038.1
XP_016867527.1
16 - 17
2673
95
XM_017012039.1
XP_016867528.1
13 - 14
1920
95
XM_017012040.1
XP_016867529.1
13 - 14
2456
95
XM_017012041.1
XP_016867530.1
15 - 16
2577
95
XM_017012042.1
XP_016867531.1
11 - 12
1497
95
XM_017012043.1
XP_016867532.1
12 - 13
1687
95
XM_017012044.1
XP_016867533.1
8 - 9
1286
95
XM_017012045.1
XP_016867534.1
8 - 9
950
95
XM_017012046.1
XP_016867535.1
12 - 13
2302
95
XM_017012047.1
XP_016867536.1
17 - 18
3285
95
XM_017012048.1
XP_016867537.1
16 - 17
3169
95
XM_017012049.1
XP_016867538.1
17 - 18
2791
95
XM_017012050.1
XP_016867539.1
16 - 17
2741
95
XM_017012051.1
XP_016867540.1
15 - 16
2645
95
XM_017012052.1
XP_016867541.1
15 - 16
2985
95
XM_017012053.1
XP_016867542.1
16 - 17
2607
95
XM_017012054.1
XP_016867543.1
15 - 16
2557
95
XM_017012055.1
XP_016867544.1
12 - 13
2340
95
XM_017012056.1
XP_016867545.1
11 - 12
3172
95
XM_017012057.1
XP_016867546.1
14 - 15
2461
95
XM_017012058.1
XP_016867547.1
10 - 11
1332
95
XM_017012059.1
XP_016867548.1
10 - 11
1454
95
XM_017012060.1
XP_016867549.1
18 - 19
3418
95
XM_017012061.1
XP_016867550.1
9 - 10
1284
95
XM_017012062.1
XP_016867551.1
8 - 9
1045
95
XM_017012063.1
XP_016867552.1
16 - 17
3150
95
XM_017012064.1
XP_016867553.1
15 - 16
3034
95
XM_017012065.1
XP_016867554.1
13 - 14
2486
95
XM_017012066.1
XP_016867555.1
11 - 12
2845
95
XM_017012067.1
XP_016867556.1
11 - 12
1348
95
XM_017012068.1
XP_016867557.1
11 - 12
2214
95
GenBank mRNA
AB000731.1
-
8 - 9
991
95
AF000017.1
-
8 - 9
985
95
AF001534.1
-
8 - 9
1016
95
AJ271366.1
-
11 - 12
1455
95
AK289511.1
-
10 - 11
1310
95
BC024285.1
-
9 - 10
1120
4590330
95
BX649028.1
-
8 - 9
1076
95
D86962.1
-
10 - 11
1748
95
U66065.1
-
8 - 9
1096
95
U69276.1
-
8 - 9
1098
95
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length between 86 and 100
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene GRB10
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of Wnt signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of phosphorylation
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
insulin-like growth factor receptor signaling pathway
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0