Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
ENST00000348066.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
ENST00000358183.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
ENST00000361571.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
ENST00000381311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
ENST00000414043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
NM_001171166.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
NM_001171167.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
NM_001171168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
NM_015099.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17013969.hg.1 |
TC17001039.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000348066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000361571.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000381311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000414043.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000572543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000574442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000574951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171167.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_015099.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000348066.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000361571.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000381311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000414043.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000572543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000574442.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
ENST00000574951.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171166.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171167.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_001171168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
NM_015099.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700173489.hg.1 |
TC1700009541.hg.1 |
Clariom D TC |