Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002036.hg.1 |
ENST00000298530.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002036.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002036.hg.1 |
TC12000158.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002037.hg.1 |
ENST00000298530.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002037.hg.1 |
ENST00000381923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002037.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002037.hg.1 |
TC12000158.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002041.hg.1 |
ENST00000298530.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002041.hg.1 |
ENST00000381923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002041.hg.1 |
ENST00000536952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002041.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002041.hg.1 |
TC12000158.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002039.hg.1 |
ENST00000298530.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002039.hg.1 |
ENST00000381923.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002039.hg.1 |
ENST00000536952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002039.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002039.hg.1 |
TC12000158.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000545924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000545924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002035.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002035.hg.1 |
TC12000158.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147698.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147700.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147702.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200147695.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147695.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147695.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200147696.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147696.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147696.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147696.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147696.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000334148.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000381923.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000545924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000545924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
ENST00000546153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_001079815.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006786.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
ENST00000298530.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
ENST00000543484.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
NM_153022.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147697.hg.1 |
TC1200006786.hg.1 |
Clariom D TC |