Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC031976.1 | - | 4 - 5 | 535 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |